Learn More
enQuireBio™ Recombinant Human Eptifibatide Protein
Recombinant Protein
Marca:  enQuireBio™ QP11802-100mg
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15998418
  • 595.00€ / 100 miligramos
Fecha estimada de envío: 01-07-2024
para ver el stock
Especificaciones
Eptifibatide Protein
Research Use Only
Recombinant Protein
E. coli
Eptifibatide was lyophilized with no additives.
100 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.