missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ELAV-like protein 1
A DNA sequence encoding the Homo sapiens (Human) ELAV-like protein 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5980-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001164666.1 | |
ELAV-like protein 1 | |
Research Use Only | |
ELAVL2 | |
Recombinant Protein | |
E. coli | |
QLSNGPTCNN TANGPTTINN NCSSPVDSGN TEDSKTNLIV NYLPQNMTQE ELKSLFGSIG EIESCKLVRD KITGQSLGYG FVNYIDPKDA EKAINTLNGL RLQTKTIKVS YARPSSASIR DANLYVSGLP KTMTQKELEQ LFSQYGRIIT SRILVDQVTG ISRGVGFIRF DKRIEAEEAI KGLNGQKPPG ATEPITVKFA NNPSQKTNQA ILSQLYQSPN RRYPGPLAQQ AQRFRLDNLL NMAYGVKRFS PMTIDGMTSL AGINIPGHPG TGWCIFVYNL APDADESILW QMFGPFGAVT NVKVIRDFNT NKCKGFGFVT MTNYDEAAMA IASLNGYRLG DRVLQVSFKT NKT | |
Greater than 90% as determined by SDS-PAGE. |
1993 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |