missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EG-VEGF / prokineticin-1 Protein
A DNA sequence encoding the Human EG-VEGF / prokineticin-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10402-ec-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_115790.1 | |
EG-VEGF / prokineticin-1 Protein | |
Research Use Only | |
PROK1 | |
The ED50 as determined by a cell proliferation assay using bovine EJG cells is less than 2.0 ug/ml, corresponding to a specific activity of > 500 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, 0.02 % Tween-20, lyophilized |
84432 | |
20 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
AVITGACERD VQCGAGTCCA ISLWLRGLRM CTPLGREGEE CHPGSHKVPF FRKRKHHTCP CLPNLLCSRF PDGRYRCSMD LKNINF | |
Greater than 98% as determined by SDS-PAGE and HPLC. |