missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EFCAB14 Protein
A DNA sequence encoding the Homo sapiens (Human) EFCAB14, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8477-ec-1mg
Additional Details : Peso : 0.01000kg
Specifications
NP_055589.1 | |
EFCAB14 Protein | |
Research Use Only | |
EFCAB14 | |
Recombinant Protein | |
E. coli | |
MKKRKELNAL IGLAGDSRRK KPKKGPSSHR LLRTEPPDSD SESSSEEEEE FGVVGNRSRF AKGDYLRCCK ICYPLCGFVI LAACVVACVG LVWMQVALKE DLDALKEKFR TMESNQKSSF QEIPKLNEEL LSKQKQLEKI ESGEMGLNKV WINITEMNKQ ISLLTSAVNH LKANVKSAAD LISLPTTVEG LQKSVASIGN TLNSVHLAVE ALQKTVDEHK KTMELLQSDM NQHFLKETPG SNQIIPSPSA TSELDNKTHS ENLKQDILYL HNSLEEVNSA LVGYQRQNDL KLEGMNETVS NLTQRVNLIE SDVVAMSKVE KKANLSFSMM GDRSATLKRQ SLDQVTNRTD TVKIQSIKKE DSSNSQVSKL REKLQLISAL TNKPESNRPP ETADEEQVES FTSKPSALPK FSQFLGDPVE KAAQLRPISL PGVSSTEDLQ DLFRKTGQDV DGKLTYQEIW TSLGSAMPEP ESLRAFDSDG DGR | |
Greater than 90% as determined by SDS-PAGE. |
9813 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |