Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase NRDP1 Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase NRDP1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8174-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001229755.1 | |
E3 ubiquitin-protein ligase NRDP1 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
10193 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
RNF41 | |
Recombinant Protein | |
E. coli | |
MGYDVTRFQG DVDEDLICPI CSGVLEEPVQ APHCEHAFCN ACITQWFSQQ QTCPVDRSVV TVAHLRPVPR IMRNMLSKLQ IACDNAVFGC SAVVRLDNLM SHLSDCEHNP KRPVTCEQGC GLEMPKDELP NHNCIKHLRS VVQQQQTRIA ELEKTSAEHK HQLAEQKRDI QLLKAYMRAI RSVNPNLQNL EETIEYNEIL EWVNSLQPAR VTRWGGMIST PDAVLQAVIK RSLVESGCPA SIVNELIENA HERSWPQGLA TLETRQMNRR YYENYVAKRI PGKQAVVVMA CENQHMGDDM VQEPGLVMIF AHGVEEI |