Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase MYLIP Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase MYLIP, was expressed in the hosts and tags indicated.
Marca:  enQuireBio™ QP7993-ye-100ug
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 16072051
  • 645.00€ / 100 microgramos
Fecha estimada de envío: 01-07-2024
para ver el stock
Especificaciones
NP_037394.2
E3 ubiquitin-protein ligase MYLIP Protein
Research Use Only
MYLIP
Recombinant Protein
Yeast
MLCYVTRPDA VLMEVEVEAK ANGEDCLNQV CRRLGIIEVD YFGLQFTGSK GESLWLNLRN RISQQMDGLA PYRLKLRVKF FVEPHLILQE QTRHIFFLHI KEALLAGHLL CSPEQAVELS ALLAQTKFGD YNQNTAKYNY EELCAKELSS ATLNSIVAKH KELEGTSQAS AEYQVLQIVS AMENYGIEWH SVRDSEGQKL LIGVGPEGIS ICKDDFSPIN RIAYPVVQMA TQSGKNVYLT VTKESGNSIV LLFKMISTRA ASGLYRAITE THAFYRCDTV TSAVMMQYSR DLKGHLASLF LNENINLGKK YVFDIKRTSK EVYDHARRAL YNAGVVDLVS RNNQSPSHSP LKSSESSMNC SSCEGLSCQQ TRVLQEKLRK LKEAMLCMVC CEEEINSTFC PCGHTVCCES CAAQLQSCPV CRSRVEHVQH VYLPTHTSLL NLTVI
Greater than 90% as determined by SDS-PAGE.
29116
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol