missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DYNC2LI1 Protein
A DNA sequence encoding the Homo sapiens (Human) DYNC2LI1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7924-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001180393.1 | |
DYNC2LI1 Protein | |
100 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
51626 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
DYNC2LI1 | |
Recombinant Protein | |
E. coli | |
MPSETLWEIA KAEVEKRGIN GSEGDGAEIA EKFVFFIGSK NGGKTTIILR CLDRDEPPKP TLALEYTYGR RAKGHNTPKD IAHFWELGGG TSLLDLISIP ITGDTLRTFS LVLVLDLSKP NDLWPTMENL LQATKSHVDK VIMKLGKTNA KAVSEMRQKI WNNMPKDHPQ DHELIDPFPV PLVIIGSKYD VFQDFESEKR KVICKTLRFV AHYYGASLMF TSKSEALLLK IRGVINQLAF GIDKSKSICV DQNKPLFITA GLDSFGQIGS PPVPENDIGK LHAHSPMELW KKVYEKLFPP KSINTLKDIK DPARDPQYAE NEVDEMRIQK DLELEQYKRS SSKSWKQIEL DS |