missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DYDC2 / DPY30 domain containing 2 Protein
A DNA sequence encoding the Homo sapiens (Human) DYDC2 / DPY30 domain containing 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7875-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001256970.1 | |
DYDC2 / DPY30 domain containing 2 Protein | |
Research Use Only | |
DYDC2 | |
Recombinant Protein | |
E. coli | |
METNYLKRCF GNCLAQALAE VAKVRPSDPI EYLAHWLYHY RKTAKAKEEN REKKIHLQEE YDSSLKEMEM TEMLKQEEYQ IQQNCEKCHK ELTSETVSTK KTIFMQEDTN PLEKEALKQE FLPGTSSLIP GMPQQVPPSE SAGQIDQNFK MPQEINYKEA FQHEVAHEMP PGSKSPF | |
Greater than 90% as determined by SDS-PAGE. |
84332 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |