missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DUSP18 Protein
A cDNA sequence encoding the DUSP18 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10577-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
DUSP18 Protein | |
Research Use Only | |
Specific activity is > 300 units/mg, and is defined as the amount of enzyme that hydrolyze 1.0 nmole of p-nitrophenyl phosphate (pNPP) per minute at pH 7.5 at 37C. | |
Human | |
His | |
The DUSP18 protein solution (0.5 mg/ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 0.1mM PMSF, 1mM DTT, 40% glycerol and 1mM EDTA. |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMTAPSC AFPVQFRQPS VSGLSQITKS LYISNGVAAN NKLMLSSNQI TMVINVSVEV VNTLYEDIQY MQVPVADSPN SRLCDFFDPI ADHIHSVEMK QGRTLLHCAA GVSRSAALCL AYLMKYHAMS LLDAHTWTKS CRPIIRPNSG FWEQLIHYEF QLFGKNTVHM VSSPVGMIPD IYEKEVRLMI PL | |
Greater than 95.0% as determined by SDS-PAGE. |