missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DHDDS Protein
A DNA sequence encoding the Homo sapiens (Human) DHDDS, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5932-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001230493.1 | |
DHDDS Protein | |
Research Use Only | |
DHDDS | |
Recombinant Protein | |
E. coli | |
MSWIKEGELS LWERFCANII KAGPMPKHIA FIMDGNRRYA KKCQVERQEG HSQGFNKLAE TLRWCLNLGI LEVTVYAFSI ENFKRSKSEV DGLMDLARQK FSRLMEEKEK LQKHGVCIRV LGDLHLLPLD LQELIAQAVQ ATKNYNKCFL NVCFAYTSRH EISNAVREMA WGVEQGLLDP SDISESLLDK CLYTNRSPHP DILIRTSGEV RLSDFLLWQT SHSCLVFQPV LWPEYTFWNL FEAILQFQMN HSVLQKARDM YAEERKRQQL ERDQATVTEQ LLREGLQASG DAQLRRTRLH KLSARREERV QGFLQALELK RADWLARLGT ASA | |
Greater than 80% as determined by SDS-PAGE. |
79947 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |