missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DENN domain-containing protein 1A Protein
A DNA sequence encoding the Homo sapiens (Human) DENN domain-containing protein 1A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7858-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Specifications
NP_065997.1 | |
DENN domain-containing protein 1A Protein | |
Research Use Only | |
DENND1A | |
Recombinant Protein | |
E. coli | |
MGSRIKQNPE TTFEVYVEVA YPRTGGTLSD PEVQRQFPED YSDQEVLQTL TKFCFPFYVD SLTVSQVGQN FTFVLTDIDS KQRFGFCRLS SGAKSCFCIL SYLPWFEVFY KLLNILADYT TKRQENQWNE LLETLHKLPI PDPGVSVHLS VHSYFTVPDT RELPSIPENR NLTEYFVAVD VNNMLHLYAS MLYERRILII CSKLSTLTAC IHGSAAMLYP MYWQHVYIPV LPPHLLDYCC APMPYLIGIH LSLMEKVRNM ALDDVVILNV DTNTLETPFD DLQSLPNDVI SSLKNRLKKV STTTGDGVAR AFLKAQAAFF GSYRNALKIE PEEPITFCEE AFVSHYRSGA MRQFLQNATQ LQLFKQFIDG RLDLLNSGEG FSDVFEEEIN MGEYAGSDKL YHQWLSTVRK GSGAILNTVK TKANPAMKTV YKFAKDHAKM GIKEVKNRLK QKDIAENGCA PTPEEQLPKT APSPLVEAKD PKLREDRRPI TVHFGQVRPP RPHVVKRPKS NIAVEGRRTS VPSPEQNTIA TPATLHILQK SITHFAAKFP TRGWTSSSH | |
Greater than 90% as determined by SDS-PAGE. |
57706 | |
200 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |