missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Delta-like protein 1
A DNA sequence encoding the Homo sapiens (Human) Delta-like protein 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8179-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_058637.1 | |
Delta-like protein 1 | |
Research Use Only | |
DLL3 | |
Recombinant Protein | |
E. coli | |
AGVFELQIHS FGPGPGPGAP RSPCSARLPC RLFFRVCLKP GLSEEAAESP CALGAALSAR GPVYTEQPGA PAPDLPLPDG LLQVPFRDAW PGTFSFIIET WREELGDQIG GPAWSLLARV AGRRRLAAGG PWARDIQRAG AWELRFSYRA RCEPPAVGTA CTRLCRPRSA PSRCGPGLRP CAPLEDECEA PLVCRAGCSP EHGFCEQPGE CRCLEGWTGP LCTVPVSTSS CLSPRGPSSA TTGCLVPGPG PCDGNPCANG GSCSETPRSF ECTCPRGFYG LRCEVSGVTC ADGPCFNGGL CVGGADPDSA YICHCPPGFQ GSNCEKRVDR CSLQPCRNGG LCLDLGHALR CRCRAGFAGP RCEHDLDDCA GRACANGGTC VEGGGAHRCS CALGFGGRDC RERADPCAAR PCAHGGRCYA HFSGLVCACA PGYMGARCEF PVHPDGASAL PAAPPGLRPG DPQRYL | |
Greater than 90% as determined by SDS-PAGE. |
10683 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |