missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DDAH1 Protein
A cDNA sequence encoding the DDAH1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11606-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
23576 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAGLGHP AAFGRATHAV VRALPESLGQ HALRSAKGEE VDVARAERQH QLYVGVLGSK LGLQVVELPA DESLPDCVFV EDVAVVCEET ALITRPGAPS RRKEVDMMKE ALEKLQLNIV EMKDENATLD GGDVLFTGRE FFVGLSKRTN QRGAEILADT FKDYAVSTVP VADGLHLKSF CSMAGPNLIA IGSSESAQKA LKIMQQMSDH RYDKLTVPDD IAANCIYLNI PNKGHVLLHR TPEEYPESAK VYEKLKDHML IPVSMSELEK VDGLLTCCSV LINKKVDS | |
Greater than 95% as determined by SDS-PAGE. |
DDAH1 Protein | |
Research Use Only | |
DDAH1 | |
Human | |
His | |
The DDAH1 protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0) 1mM DTT, 50mM NaCl and 10% glycerol. |