missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cystathionine beta-synthase Protein
A DNA sequence encoding the Homo sapiens (Human) Cystathionine beta-synthase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5767-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000062.1 | |
Cystathionine beta-synthase Protein | |
Research Use Only | |
CBS | |
Recombinant Protein | |
E. coli | |
PSETPQAEVG PTGCPHRSGP HSAKGSLEKG SPEDKEAKEP LWIRPDAPSR CTWQLGRPAS ESPHHHTAPA KSPKILPDIL KKIGDTPMVR INKIGKKFGL KCELLAKCEF FNAGGSVKDR ISLRMIEDAE RDGTLKPGDT IIEPTSGNTG IGLALAAAVR GYRCIIVMPE KMSSEKVDVL RALGAEIVRT PTNARFDSPE SHVGVAWRLK NEIPNSHILD QYRNASNPLA HYDTTADEIL QQCDGKLDML VASVGTGGTI TGIARKLKEK CPGCRIIGVD PEGSILAEPE ELNQTEQTTY EVEGIGYDFI PTVLDRTVVD KWFKSNDEEA FTFARMLIAQ EGLLCGGSAG STVAVAVKAA QELQEGQRCV VILPDSVRNY MTKFLSDRWM LQKGFLKEED LTEKKPWWWH LRVQELGLSA PLTVLPTITC GHTIEILREK GFDQAPVVDE AGVILGMVTL GNMLSSLLAG KVQPSDQVGK VIYKQFKQIR LTDTLGRLSH ILEMDHFALV VHEQIQYHST GKSSQRQMVF GVVTAIDLLN FVAAQERDQK | |
Greater than 90% as determined by SDS-PAGE. |
102724560 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |