missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CYB5R1 Protein
A cDNA sequence encoding the CYB5R1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11573-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CYB5R1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSLVRRSRR PQVTLLDPNE KYLLRLLDKT TVSHNTKRFR FALPTAHHTL GLPVGKHIYL STRIDGSLVI RPYTPVTSDE DQGYVDLVIK VYLKGVHPKF PEGGKMSQYL DSLKVGDVVE FRGPSGLLTY TGKGHFNIQP NKKSPPEPRV AKKLGMIAGG TGITPMLQLI RAILKVPEDP TQCFLLFANQ TEKDIILRED LEELQARYPN RFKLWFTLDH PPKDWAYSKG FVTADMIREH LPAPGDDVLV LLCGPPPMVQ LACHPNLDKL GYSQKMRFTY | |
Greater than 85% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CYB5R1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |