missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CXCL4 / PF4 Protein
A DNA sequence encoding the Human CXCL4 / PF4 Protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP1018-ec-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002610.1 | |
CXCL4 / PF4 Protein | |
Research Use Only | |
PF4 | |
The biological activity determined by a chemotaxis bioassay using human fibroblasts is in a concentration of 1.0-10 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
5196 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
EAEEDGDLQC LCVKTTSQVR PRHITSLEVI KAGPHCPTAQ LIATLKNGRK ICLDLQAPLY KKIIKKLLES | |
Greater than 97% as determined by SDS-PAGE and HPLC. |