missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CXCL12 / SDF-1 Protein
A DNA sequence encoding the Human CXCL12 / SDF-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10216-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000600.1 | |
CXCL12 / SDF-1 Protein | |
Research Use Only | |
CXCL12 | |
The biological activity determined by a chemotaxis bioassay using PHA and rHuIL-2 activated human peripheral blood T-lymphocytes is in a concentration range of 20-80 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
6387 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNK | |
Greater than 97% as determined by SDS-PAGE and HPLC. |