missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CRP Protein
A cDNA sequence encoding the CRP was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11511-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
1401 | |
Greater than 96.0%. | |
Research Use Only | |
CRP | |
Human | |
Untagged | |
The protein solution is in 20mM TRIS buffer pH 8.0 containing 0.28M NaCl, 0.09% NaN3 and 5mM CaCl2. |
CRP Protein | |
200 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Native Protein | |
MEKLLCFLVL TSLSHAFGQT DMSRKAFVFP KESDTSYVSL KAPLTKPLKA FTVCLHFYTE LSSTRGYSIF SYATKRQDNE ILIFWSKDIG YSFTVGGSEI LFEVPEVTVA PVHICTSWES ASGIVEFWVD GKPRVRKSLK KGYTVGAEAS IILGQEQDSF GGNFEGSQSL VGDIGNVNMW DFVLSPDEIN TIYLGGPFSP NVLNWRALKY EVQGEVFTKP QLWP |