missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CRP Protein

A cDNA sequence encoding the CRP was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11511-10mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15937488

Cantidad Precio
1 1010.00€ / 10 miligramos
4 941.52€ / 10 miligramos
Fecha estimada de envío
21-10-2020
Añadir a la cesta


Especificaciones

Especificaciones

1401
Greater than 96.0%.
Research Use Only
CRP
Human
Untagged
The protein solution is in 20mM TRIS buffer pH 8.0 containing 0.28M NaCl, 0.09% NaN3 and 5mM CaCl2.
CRP Protein
10 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Native Protein
MEKLLCFLVL TSLSHAFGQT DMSRKAFVFP KESDTSYVSL KAPLTKPLKA FTVCLHFYTE LSSTRGYSIF SYATKRQDNE ILIFWSKDIG YSFTVGGSEI LFEVPEVTVA PVHICTSWES ASGIVEFWVD GKPRVRKSLK KGYTVGAEAS IILGQEQDSF GGNFEGSQSL VGDIGNVNMW DFVLSPDEIN TIYLGGPFSP NVLNWRALKY EVQGEVFTKP QLWP
Videos
Documentos