missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human COX7A2L, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) COX7A2L, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5855-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001305965.1 | |
COX7A2L, mitochondrial Protein | |
Research Use Only | |
COX7A2L | |
Recombinant Protein | |
E. coli | |
MYYKFSGFTQ KLAGAWASEA YSPQGLKPVV STEAPPIIFA TPTKLTSDST VYDYAGKNKV PELQKFFQKA DGVPVYLKRG LPDQMLYRTT MALTVGGTIY CLIALYMASQ PKNK | |
Greater than 80% as determined by SDS-PAGE. |
9167 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |