missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CNTF Protein
A DNA sequence encoding the Human CNTF, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10413-ec-250ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000605.1 | |
CNTF Protein | |
Research Use Only | |
CNTF | |
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 150 ng/ml, corresponding to a specific activity of > 6.7 M 103 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
1270 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MAFTEHSPLT PHRRDLCSRS IWLARKIRSD LTALTESYVK HQGLNKNINL DSADGMPVAS TDQWSELTEA ERLQENLQAY RTFHVLLARL LEDQQVHFTP TEGDFHQAIH TLLLQVAAFA YQIEELMILL EYKIPRNEAD GMPINVGDGG LFEKKLWGLK VLQELSQWTV RSIHDLRFIS SHQTGIPARG SHYIANNKKM | |
Greater than 97% as determined by SDS-PAGE and HPLC. |