missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CKMT1 Protein
A cDNA sequence encoding the CKMT1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10555-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
1158 | |
Greater than 95.0% as determined bya) Analysis by RP-HPLCb) Analysis by SDS-PAGE | |
Research Use Only | |
CKM | |
Recombinant Protein | |
Pichia Pastoris | |
MPFGNTHNKF KLNYKPEEEY PDLSKHNNHM AKVLTLELYK KLRDKETPSG FTVDDVIQTG VDNPGHPFIM TVGCVAGDEE SYEVFKELFD PIISDRHGGY KPTDKHKTDL NHENLKGGDD LDPNYVLSSR VRTGRSIKGY TLPPHCSRGE RRAVEKLSVE ALNSLTGEFK GKYYPLKSMT EKEQQQLIDD HFLFDKPVSP LLLASGMARD WPDARGIWHN DNKSFLVWVN EEDHLRVISM EKGGNMKEVF RRFCVGLQKI EEIFKKAGHP FMWNQHLGYV LTCPSNLGTG LRGGVHVKLA HLSKHPKFEE ILTRLRLQKR GTGGVDTAAV GSVFDVSNAD RLGSSEVEQV QLVVDGVKLM VEMEKKLEKG QSIDDMIPAQ K |
CKMT1 Protein | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The biological activity measured by the enzymatic activity of Creatine phosphokinase procedure No. 45-UV, 1IU-1 micromole creatine phosphate was 537IU/mg at 37 degrees celsius corresponding to a Specific Activity of 1,863ng/ml. | |
Human | |
Untagged | |
CKMT1 contains 0.01M Tris-HCl, 0.075M NaCl, 10mM beta-mercaptoethanol, 50% glycerol, 0.1% sodium azide, pH 7.2. |