missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CIB2 Protein
A cDNA sequence encoding the CIB2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11429-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
10518 | |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGNKQTIFTE EQLDNYQDCT FFNKKDILKL HSRFYELAPN LVPMDYRKSP IVHVPMSLII QMPELRENPF KERIVAAFSE DGEGNLTFND FVDMFSVLCE SAPRELKANY AFKIYDFNTD NFICKEDLEL TLARLTKSEL DEEEVVLVCD KVIEEADLDG DGKLGFADFE DMIAKAPDFL STFHIRI | |
Greater than 90.0% as determined by SDS-PAGE. |
CIB2 Protein | |
Research Use Only | |
CIB2 | |
Human | |
His | |
The CIB2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 2mM DTT, 100mM NaCl and 1mM PMSF. |