missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CEACAM1 / CD66a Protein
A DNA sequence encoding the Homo sapiens (Human) CEACAM1 / CD66a, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5811-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001020083.1 | |
CEACAM1 / CD66a Protein | |
Research Use Only | |
CEACAM1 | |
Recombinant Protein | |
E. coli | |
QLTTESMPFN VAEGKEVLLL VHNLPQQLFG YSWYKGERVD GNRQIVGYAI GTQQATPGPA NSGRETIYPN ASLLIQNVTQ NDTGFYTLQV IKSDLVNEEA TGQFHVYPEL PKPSISSNNS NPVEDKDAVA FTCEPETQDT TYLWWINNQS LPVSPRLQLS NGNRTLTLLS VTRNDTGPYE CEIQNPVSAN RSDPVTLNVT YGPDTPTISP SDTYYRPGAN LSLSCYAASN PPAQYSWLIN GTFQQSTQEL FIPNITVNNS GSYTCHANNS VTGCNRTTVK TIIVTELSPV VAKPQIKASK TTVTGDKDSV NLTCSTNDTG ISIRWFFKNQ SLPSSERMKL SQGNTTLSIN PVKREDAGTY WCEVFNPISK NQSDPIMLNV NYNALPQENG LSPG | |
Greater than 90% as determined by SDS-PAGE. |
634 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |