missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD58 Protein
A cDNA sequence encoding the CD58 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11340-HIS-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
965 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSFSQQIYG VVYGNVTFHV PSNVPLKEVL WKKQKDKVAE LENSEFRAFS SFKNRVYLDT VSGSLTIYNL TSSDEDEYEM ESPNITDTMK FFLYVLESLP SPTLTCALTN GSIEVQCMIP EHYNSHRGLI MYSWDCPMEQ CKRNSTSIYF KMENDLPQKI QCTLSNPLFN TTSSIILTTC IPSSGHSRHR | |
Greater than 90.0% as determined by SDS-PAGE. |
CD58 Protein | |
Research Use Only | |
CD58 | |
Human | |
His | |
CD58 protein solution (1 mg/ml) containing 20mM Phosphate buffer and 10% glycerol. |