missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD23 / FCER2 / FCER2A Protein
A DNA sequence encoding the Homo sapiens (Human) CD23 / FCER2 / FCER2A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6029-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001193948.2 | |
CD23 / FCER2 / FCER2A Protein | |
Research Use Only | |
FCER2 | |
Recombinant Protein | |
E. coli | |
DTTQSLKQLE ERAARNVSQV SKNLESHHGD QMAQKSQSTQ ISQELEELRA EQQRLKSQDL ELSWNLNGLQ ADLSSFKSQE LNERNEASDL LERLREEVTK LRMELQVSSG FVCNTCPEKW INFQRKCYYF GKGTKQWVHA RYACDDMEGQ LVSIHSPEEQ DFLTKHASHT GSWIGLRNLD LKGEFIWVDG SHVDYSNWAP GEPTSRSQGE DCVMMRGSGR WNDAFCDRKL GAWVCDRLAT CTPPASEGSA ESMGPDSRPD PDGRLPTPSA PLHS | |
Greater than 90% as determined by SDS-PAGE. |
2208 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |