missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD133 / PROM1 / Prominin 1 Protein
A DNA sequence encoding the Homo sapiens (Human) CD133 / PROM1 / Prominin 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9348-ye-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001139319.1 | |
CD133 / PROM1 / Prominin 1 Protein | |
Research Use Only | |
PROM1 | |
Recombinant Protein | |
Yeast | |
GANVEKLICE PYTSKELFRV LDTPYLLNED WEYYLSGKLF NKSKMKLTFE QVYSDCKKNR GTYGTLHLQN SFNISEHLNI NEHTGSISSE LESLKVNLNI FLLGAAGRKN LQDFAACGID RMNYDSYLAQ TGKSPAGVNL LSFAYDLEAK ANSLPPGNLR NSLKRDAQTI KTIHQQRVLP IEQSLSTLYQ SVKILQRTGN GLLERVTRIL ASLDFAQNFI TNNTSSVIIE ETKKYGRTII GYFEHYLQWI EFSISEKVAS CKPVATALDT AVDVFLCSYI IDPLN | |
Greater than 90% as determined by SDS-PAGE. |
8842 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |