missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL20 / MIP-3 alpha Protein
A DNA sequence encoding the Homo sapiens (Human) CCL20 / MIP-3 alpha, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8520-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001123518.1 | |
CCL20 / MIP-3 alpha Protein | |
Research Use Only | |
CCL20 | |
Recombinant Protein | |
E. coli | |
ASNFDCCLGY TDRILHPKFI VGFTRQLANE GCDINAIIFH TKKKLSVCAN PKQTWVKYIV RLLSKKVKN | |
Greater than 90% as determined by SDS-PAGE. |
6364 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |