missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL11 / Eotaxin Protein
A DNA sequence encoding the Homo sapiens (Human) CCL11 / Eotaxin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5770-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002977.1 | |
CCL11 / Eotaxin Protein | |
Research Use Only | |
CCL11 | |
Recombinant Protein | |
E. coli | |
GPASVPTTCC FNLANRKIPL QRLESYRRIT SGKCPQKAVI FKTKLAKDIC ADPKKKWVQD SMKYLDQKSP TPKP | |
Greater than 80% as determined by SDS-PAGE. |
6356 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |