missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CALML3 Protein
A DNA sequence encoding the Homo sapiens (Human) CALML3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5748-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_005176.1 | |
CALML3 Protein | |
Research Use Only | |
CALML3 | |
Recombinant Protein | |
E. coli | |
MADQLTEEQV TEFKEAFSLF DKDGDGCITT RELGTVMRSL GQNPTEAELR DMMSEIDRDG NGTVDFPEFL GMMARKMKDT DNEEEIREAF RVFDKDGNGF VSAAELRHVM TRLGEKLSDE EVDEMIRAAD TDGDGQVNYE EFVRVLVSK | |
Greater than 80% as determined by SDS-PAGE. |
810 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |