missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BNIP3L Protein
A DNA sequence encoding the Homo sapiens (Human) BNIP3L, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9010-ye-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001317420.1 | |
BNIP3L Protein | |
Research Use Only | |
BNIP3L | |
Recombinant Protein | |
Yeast | |
MSSHLVEPPP PLHNNNNNCE ENEQSLPPPA GLNSSWVELP MNSSNGNDNG NGKNGGLEHV PSSSSIHNGD MEKILLDAQH ESGQSSSRGS SHCDSPSPQE DGQIMFDVEM HTSRDHSSQS EEEVVEGEKE VEALKKSADW VSDWSSRPEN IPPKEFHFRH PKRSVSLSMR KSGAMKKGGI FSAEFLKVFI PSLFLSHVLA LGLGIYIGKR LSTPSASTY | |
Greater than 90% as determined by SDS-PAGE. |
665 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |