missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BLOC1S5 Protein
A cDNA sequence encoding the BLOC1S5 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11169-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
63915 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSGGGTE TPVGCEAAPG GGSKKRDSLG TAGSAHLIIK DLGEIHSRLL DHRPVIQGET RYFVKEFEEK RGLREMRVLE NLKNMIHETN EHTLPKCRDT MRDSLSQVLQ RLQAANDSVC RLQQREQERK KIHSDHLVAS EKQHMLQWDN FMKEQPNKRA EVDEEHRKAM ERLKEQYAEM EKDLAKFSTF | |
Greater than 90.0% as determined by SDS-PAGE. |
BLOC1S5 Protein | |
Research Use Only | |
BLOC1S5 | |
Human | |
His | |
BLOC1S5 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 40% glycerol, 2mM DTT, 0.1mM PMSF and 1mM EDTA. |