missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BLMH Protein
A cDNA sequence encoding the BLMH was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10535-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
642 | |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity: > 1,000 pmole/min/ug. Measured by the hydrolysis of Met-AMC at pH 7.5, at 37C. | |
Human | |
His | |
The BLMH protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl Buffer (pH 8.0) and 10% Glycerol. |
BLMH Protein | |
Research Use Only | |
BLMH | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSSSGLNSEK VAALIQKLNS DPQFVLAQNV GTTHDLLDIC LKRATVQRAQ HVFQHAVPQE GKPITNQKSS GRCWIFSCLN VMRLPFMKKL NIEEFEFSQS YLFFWDKVER CYFFLSAFVD TAQRKEPEDG RLVQFLLMNP ANDGGQWDML VNIVEKYGVI PKKCFPESYT TEATRRMNDI LNHKMREFCI RLRNLVHSGA TKGEISATQD VMMEEIFRVV CICLGNPPET FTWEYRDKDK NYQKIGPITP LEFYREHVKP LFNMEDKICL VNDPRPQHKY NKLYTVEYLS NMVGGRKTLY NNQPIDFLKK MVAASIKDGE AVWFGCDVGK HFNSKLGLSD MNLYDHELVF GVSLKNMNKA ERLTFGESLM THAMTFTAVS EKDDQDGAFT KWRVENSWGE DHGHKGYLCM TDEWFSEYVY EVVVDRKHVP EEVLAVLEQE PIILPAWDPM GALAE | |
Greater than 90% as determined by SDS-PAGE. |