missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BIN2 Protein
A cDNA sequence encoding the BIN2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11163-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
BIN2 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPMRGMPGA RTSSSGASEN HRARGQGGGP QGVGRMAEGK AGGAAGLFAK QVQKKFSRAQ EKVLQKLGKA VETKDERFEQ SASNFYQQQA EGHKLYKDLK NFLSAVKVMH ESSKRVSETL QEIYSSEWDG HEELKAIVWN NDLLWEDYEE KLADQAVRTM EIYVAQFSEI KERIAKRGRK LVDYDSARHH LEAVQNAKKK DEAKTAKAEE EFNKAQTVFE DLNQELLEEL PILYNSRIGC YVTIFQNISN LRDVFYREMS KLNHNLYEVM SKLEKQHSNK VFVVKGLSSS SRRSLVISPP VRTATVSSPL TSPTSPSTLS LKSESESVSA TEDLAPDAAQ GEDNSEIKEL LEEEEIEKEG SEASSSEEDE PLPACNGPAQ AQPSPTTERA KSQEEVLPSS TTPSPGGALS PSGQPSSSAT EVVLRTRTAS EGSEQPKKRA SIQRTSAPPS RPPPPRATAS PRPSSGNIPS SPTASGGGSP TSPRASLGTG TASPRTSLEV SPNPEPPEKP VRTPEAKENE NIHNQNPEEL CTSPTLMTSQ VASEPGEAKK MEDKEKDNKL ISANSSEGQD QLQVSMVPEN NNLTAPEPQE EVSTSENPQL HHHHHH | |
Greater than 80.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
BIN2 protein solution (0.25 mg/ml) contains Phosphate buffered saline (pH 7.4), 20% glycerol, 1mM DTT and 0.1mM PMSF. |