missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BCL2L1 / Bcl-XL Protein
A DNA sequence encoding the Homo sapiens (Human) BCL2L1 / Bcl-XL, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5711-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001182.1 | |
BCL2L1 / Bcl-XL Protein | |
Research Use Only | |
BCL2L1 | |
Recombinant Protein | |
E. coli | |
MSQSNRELVV DFLSYKLSQK GYSWSQFSDV EENRTEAPEG TESEMETPSA INGNPSWHLA DSPAVNGATG HSSSLDAREV IPMAAVKQAL REAGDEFELR YRRAFSDLTS QLHITPGTAY QSFEQVVNEL FRDGVNWGRI VAFFSFGGAL CVESVDKEMQ VLVSRIAAWM ATYLNDHLEP WIQENGGWDT FVELYGNNAA AESRKGQER | |
Greater than 90% as determined by SDS-PAGE. |
598 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |