missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Bcl 2 (1-206 a. a. ) Protein
A cDNA sequence encoding the Bcl 2 (1-206 a. a. ) was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10490-HIS-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Bcl 2 (1-206 a. a. ) Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MANUAL | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The protein contains 10mM Tris-HCl pH 8.0, 1mM EDTA and 250mM NaCl. |