missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BAG family molecular chaperone regulator 5 Protein
A DNA sequence encoding the Homo sapiens (Human) BAG family molecular chaperone regulator 5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8234-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001015048.1 | |
BAG family molecular chaperone regulator 5 Protein | |
Research Use Only | |
BAG5 | |
Recombinant Protein | |
E. coli | |
MDMGNQHPSI SRLQEIQKEV KSVEQQVIGF SGLSDDKNYK KLERILTKQL FEIDSVDTEG KGDIQQARKR AAQETERLLK ELEQNANHPH RIEIQNIFEE AQSLVREKIV PFYNGGNCVT DEFEEGIQDI ILRLTHVKTG GKISLRKARY HTLTKICAVQ EIIEDCMKKQ PSLPLSEDAH PSVAKINFVM CEVNKARGVL IALLMGVNNN ETCRHLSCVL SGLIADLDAL DVCGRTEIRN YRREVVEDIN KLLKYLDLEE EADTTKAFDL RQNHSILKIE KVLKRMREIK NELLQAQNPS ELYLSSKTEL QGLIGQLDEV SLEKNPCIRE ARRRAVIEVQ TLITYIDLKE ALEKRKLFAC EEHPSHKAVW NVLGNLSEIQ GEVLSFDGNR TDKNYIRLEE LLTKQLLALD AVDPQGEEKC KAARKQAVRL AQNILSYLDL KSDEWEY | |
Greater than 90% as determined by SDS-PAGE. |
9529 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |