missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BAFFR / TNFRSF13C Protein
A DNA sequence encoding the Human BAFFR / TNFRSF13C, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10356-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_443177.1 | |
BAFFR / TNFRSF13C Protein | |
Research Use Only | |
TNFRSF13C | |
The ED50 as determined by its ability to block BAFF induced mouse splenocyte survival is 1.0-5.0 ug/ml in the presence of 1.0 ug/ml of rHuBAFF. | |
Human | |
Untagged | |
0.2 um filtered 20 mM PBS, pH 8.0, 500 mM NaCl, lyophilized |
115650 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MRRGPRSLRG RDAPAPTPCV PAECFDLLVR HCVACGLLRT PRPKPAGASS PAPRTALQPQ ESVGAGAGEA ALPLPG | |
Greater than 95% as determined by SDS-PAGE. |