missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATF3 Protein
A cDNA sequence encoding the ATF3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11098-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
467 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MMLQHPGQVS ASEVSASAIV PCLSPPGSLV FEDFANLTPF VKEELRFAIQ NKHLCHRMSS ALESVTVSDR PLGVSITKAE VAPEEDERKK RRRERNKIAA AKCRNKKKEK TECLQKESEK LESVNAELKA QIEELKNEKQ HLIYMLNLHR PTCIVRAQNG RTPEDERNLF IQQIKEGTLQ S | |
Greater than 90.0% as determined by SDS-PAGE. |
ATF3 Protein | |
Research Use Only | |
ATF3 | |
Human | |
Untagged | |
ATF3 protein (1 mg/ml) is supplied in 20mM Tris-HCl, pH 8.0, 2M Urea and 20% Glycerol. |