missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Arylsulfatase G Protein
A DNA sequence encoding the Homo sapiens (Human) Arylsulfatase G, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5680-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001254656.1 | |
Arylsulfatase G Protein | |
Research Use Only | |
ARSG | |
Recombinant Protein | |
E. coli | |
GFLYPLVDFC ISGKTRGQKP NFVIILADDM GWGDLGANWA ETKDTANLDK MASEGMRFVD FHAAASTCSP SRASLLTGRL GLRNGVTRNF AVTSVGGLPL NETTLAEVLQ QAGYVTGIIG KWHLGHHGSY HPNFRGFDYY FGIPYSHDMG CTDTPGYNHP PCPACPQGDG PSRNLQRDCY TDVALPLYEN LNIVEQPVNL SSLAQKYAEK ATQFIQRAST SGRPFLLYVA LAHMHVPLPV TQLPAAPRGR SLYGAGLWEM DSLVGQIKDK VDHTVKENTF LWFTGDNGPW AQKCELAGSV GPFTGFWQTR QGGSPAKQTT WEGGHRVPAL AYWPGRVPVN VTSTALLSVL DIFPTVVALA QASLPQGRRF DGVDVSEVLF GRSQPGHRVL FHPNSGAAGE FGALQTVRLE RYKAFYITGG ARACDGSTGP ELQHKFPLIF NLEDDTAEAV PLERGGAEYQ AVLPEVRKVL ADVLQDIAND NISSADYTQD PSVTPCCNPY QIACRCQAA | |
Greater than 90% as determined by SDS-PAGE. |
22901 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |