missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Apolipoprotein A-IV Protein
A DNA sequence encoding the Homo sapiens (Human) Apolipoprotein A-IV, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5661-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Apolipoprotein A-IV Protein | |
Research Use Only | |
APOA4 | |
Recombinant Protein | |
E. coli | |
EVSADQVATV MWDYFSQLSN NAKEAVEHLQ KSELTQQLNA LFQDKLGEVN TYAGDLQKKL VPFATELHER LAKDSEKLKE EIGKELEELR ARLLPHANEV SQKIGDNLRE LQQRLEPYAD QLRTQVNTQA EQLRRQLTPY AQRMERVLRE NADSLQASLR PHADELKAKI DQNVEELKGR LTPYADEFKV KIDQTVEELR RSLAPYAQDT QEKLNHQLEG LTFQMKKNAE ELKARISASA EELRQRLAPL AEDVRGNLRG NTEGLQKSLA ELGGHLDQQV EEFRRRVEPY GENFNKALVQ QMEQLRQKLG PHAGDVEGHL SFLEKDLRDK VNSFFSTFKE KESQDKTLSL PELEQQQEQQ QEQQQEQVQM LAPLES | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |