missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human APOL4 Protein
A cDNA sequence encoding the APOL4 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11055-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
80832 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGSWVQL ITSVGVQQNH PGWTVAGQFQ EKKRFTEEVI EYFQKKVSPV HLKILLTSDE AWKRFVRVAE LPREEADALY EALKNLTPYV AIEDKDMQQK EQQFREWFLK EFPQIRWKIQ ESIERLRVIA NEIEKVHRGC VIANVVSGST GILSVIGVML APFTAGLSLS ITAAGVGLGI ASATAGIASS IVENTYTRSA ELTASRLTAT STDQLEALRD ILRDITPNVL SFALDFDEAT KMIANDVHTL RRSKATVGRP LIAWRYVPIN VVETLRTRGA PTRIVRKVAR NLGKATSGVL VVLDVVNLVQ DSLDLHKGAK SESAESLRQW AQELEENLNE LTHIHQSLKA G | |
Greater than 85.0% as determined by SDS-PAGE. |
APOL4 Protein | |
Research Use Only | |
APOL4 | |
Human | |
His | |
APOH protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 10% glycerol and 0.4M Urea. |