missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ANGPTL4 Protein
A DNA sequence encoding the Homo sapiens (Human) ANGPTL4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8682-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001034756.1 | |
ANGPTL4 Protein | |
Research Use Only | |
ANGPTL4 | |
Recombinant Protein | |
E. coli | |
VQSKSPRFAS WDEMNVLAHG LLQLGQGLRE HAERTRSQLS ALERRLSACG SACQGTEGST DLPLAPESRV DPEVLHSLQT QLKAQNSRIQ QLFHKVAQQQ RHLEKQHLRI QHLQSQFGLL DHKHLDHEVA KPARRKRLPE MAQPVDPAHN VSRLHRLPRD CQELFQVGER QSGLFEIQPQ GSPPFLVNCK MTSDGGWTVI QRRHDGSVDF NRPWEAYKAG FGDPHGEFWL GLEKVHSITG DRNSRLAVQL RDWDGNAELL QFSVHLGGED TAYSLQLTAP VAGQLGATTV PPSGLSVPFS TWDQDHDLRR DKNCAKSLSG GWWFGTCSHS NLNGQYFRSI PQQRQKLKKG IFWKTWRGRY YPLQATTMLI QPMAAE | |
Greater than 90% as determined by SDS-PAGE. |
51129 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |