missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ALDOC Protein
A cDNA sequence encoding the ALDOC was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10517-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
230 | |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 6 units/mg, one unit will convert 1.0 umol of fructose 1,6-diphosphate to dihydroxyacetone phosphate and glyceraldehydes 3- phosphate per minute at pH 7.5 at 37C. | |
Human | |
Untagged | |
The ALDOC solution (1 mg/ml) contains 20% glycerol, 20mM Tris-HCl buffer (pH 8.0) , 2mM DTT and 0.1M NaCl. |
ALDOC Protein | |
Research Use Only | |
ALDOC | |
Recombinant Protein | |
E. coli | |
MPHSYPALSA EQKKELSDIA LRIVAPGKGI LAADESVGSM AKRLSQIGVE NTEENRRLYR QVLFSADDRV KKCIGGVIFF HETLYQKDDN GVPFVRTIQD KGIVVGIKVD KGVVPLAGTD GETTTQGLDG LSERCAQYKK DGADFAKWRC VLKISERTPS ALAILENANV LARYASICQQ NGIVPIVEPE ILPDGDHDLK RCQYVTEKVL AAVYKALSDH HVYLEGTLLK PNMVTPGHAC PIKYTPEEIA MATVTALRRT VPPAVPGVTF LSGGQSEEEA SFNLNAINRC PLPRPWALTF SYGRALQASA LNAWRGQRDN AGAATEEFIK RAEVNGLAAQ GKYEGSGEDG GAAAQSLYIA NHAY | |
Greater than 90.0% as determined by SDS-PAGE. |