missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Aldo-keto reductase family 1 member C3 Protein
A DNA sequence encoding the Homo sapiens (Human) Aldo-keto reductase family 1 member C3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5638-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001240837.1 | |
Aldo-keto reductase family 1 member C3 Protein | |
Research Use Only | |
AKR1C3 | |
Recombinant Protein | |
E. coli | |
MDSKHQCVKL NDGHFMPVLG FGTYAPPEVP RSKALEVTKL AIEAGFRHID SAHLYNNEEQ VGLAIRSKIA DGSVKREDIF YTSKLWSTFH RPELVRPALE NSLKKAQLDY VDLYLIHSPM SLKPGEELSP TDENGKVIFD IVDLCTTWEA MEKCKDAGLA KSIGVSNFNR RQLEMILNKP GLKYKPVCNQ VECHPYFNRS KLLDFCKSKD IVLVAYSALG SQRDKRWVDP NSPVLLEDPV LCALAKKHKR TPALIALRYQ LQRGVVVLAK SYNEQRIRQN VQVFEFQLTA EDMKAIDGLD RNLHYFNSDS FASHPNYPYS DEY | |
Greater than 80% as determined by SDS-PAGE. |
8644 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |