missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ALDH5A1, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) ALDH5A1, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5645-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001071.1 | |
ALDH5A1, mitochondrial Protein | |
Research Use Only | |
ALDH5A1 | |
Recombinant Protein | |
E. coli | |
AGRLAGLSAA LLRTDSFVGG RWLPAAATFP VQDPASGAAL GMVADCGVRE ARAAVRAAYE AFCRWREVSA KERSSLLRKW YNLMIQNKDD LARIITAESG KPLKEAHGEI LYSAFFLEWF SEEARRVYGD IIHTPAKDRR ALVLKQPIGV AAVITPWNFP SAMITRKVGA ALAAGCTVVV KPAEDTPFSA LALAELASQA GIPSGVYNVI PCSRKNAKEV GEAICTDPLV SKISFTGSTT TGKILLHHAA NSVKRVSMEL GGLAPFIVFD SANVDMS-QA VAGAMASKFR NTGQTCVCSN QFLVQRGIHD AFVKAFAEAM KKNLRVGNGF EEGTTQGPLI NEKAVEKVEK QVNDAVSKGA TVVTGGKRHQ LGKNFFEPTL LCNVTQDMLC THEETFGPLA PVIKFDTEEE AIAIANAADV GLAGYFYSQD PAQIWRVAEQ LEVGMVGVNE GLISSVECPF GGVKQSGLGR EGSKYGIDEY LELKYVCYGG L | |
Greater than 90% as determined by SDS-PAGE. |
7915 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |