missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Alcohol dehydrogenase 1B Protein
A DNA sequence encoding the Homo sapiens (Human) Alcohol dehydrogenase 1B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5613-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000659.2 | |
Alcohol dehydrogenase 1B Protein | |
Research Use Only | |
ADH1B | |
Recombinant Protein | |
E. coli | |
STAGKVIKCK AAVLWEVKKP FSIEDVEVAP PKAYEVRIKM VAVGICRTDD HVVSGNLVTP LPVILGHEAA GIVESVGEGV TTVKPGDKVI PLFTPQCGKC RVCKNPESNY CLKNDLGNPR GTLQDGTRRF TCRGKPIHHF LGTSTFSQYT VVDENAVAKI DAASPLEKVC LIGCGFSTGY GSAVNVAKVT PGSTCAVFGL GGVGLSAVMG CKAAGAARII AVDINKDKFA KAKELGATEC INPQDYKKPI QEVLKEMTDG GVDFSFEVIG RLDTMMASLL CCHEACGTSV IVGVPPASQN LSINPMLLLT GRTWKGAVYG GFKSKEGIPK LVADFMAKKF SLDALITHVL PFEKINEGFD LLHSGKSIRT VLTF | |
Greater than 90% as determined by SDS-PAGE. |
125 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |