missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AK1 / Adenylate kinase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) AK1 / Adenylate kinase 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5634-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000467.1 | |
AK1 / Adenylate kinase 1 Protein | |
Research Use Only | |
AK1 | |
Recombinant Protein | |
E. coli | |
MEEKLKKTKI IFVVGGPGSG KGTQCEKIVQ KYGYTHLSTG DLLRSEVSSG SARGKKLSEI MEKGQLVPLE TVLDMLRDAM VAKVNTSKGF LIDGYPREVQ QGEEFERRIG QPTLLLYVDA GPETMTQRLL KRGETSGRVD DNEETIKKRL ETYYKATEPV IAFYEKRGIV RKVNAEGSVD SVFSQVCTHL DALK | |
Greater than 90% as determined by SDS-PAGE. |
203 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |