missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AIDA Protein
A cDNA sequence encoding the AIDA was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10986-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
64853 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSEVTRS LLQRWGASFR RGADFDSWGQ LVEAIDEYQI LARHLQKEAQ AQHNNSEFTE EQKKTIGKIA TCLELRSAAL QSTQSQEEFK LEDLKKLEPI LKNILTYNKE FPFDVQPVPL RRILAPGEEE NLEFEEDEEE GGAGAGSPDS FPARVPGTLL PRLPSEPGMT LLTIRIEKIG LKDAGQCIDP YITVSVKDLN GIDLTPVQDT PVASRKEDTY VHFNVDIELQ KHVEKLTKGA AIFFEFKHYK PKKRFTSTKC FAFMEMDEIK PGPIVIELYK KPTDFKRKKL QLLTKKPLYL HLHQTLHKE | |
Greater than 90% as determined by SDS-PAGE. |
AIDA Protein | |
Research Use Only | |
AIDA | |
Human | |
His | |
AIDA protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.2M NaCl and 1mM DTT. |