missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Adenosylhomocysteinase Protein
A DNA sequence encoding the Homo sapiens (Human) Adenosylhomocysteinase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5629-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000678.1 | |
Adenosylhomocysteinase Protein | |
Research Use Only | |
AHCY | |
Recombinant Protein | |
E. coli | |
SDKLPYKVAD IGLAAWGRKA LDIAENEMPG LMRMRERYSA SKPLKGARIA GCLHMTVETA VLIETLVTLG AEVQWSSCNI FSTQDHAAAA IAKAGIPVYA WKGETDEEYL WCIEQTLYFK DGPLNMILDD GGDLTNLIHT KYPQLLPGIR GISEETTTGV HNLYKMMANG ILKVPAINVN DSVTKSKFDN LYGCRESLID GIKRATDVMI AGKVAVVAGY GDVGKGCAQA LRGFGARVII TEIDPINALQ AAMEGYEVTT MDEACQEGNI FVTTTGCIDI ILGRHFEQMK DDAIVCNIGH FDVEIDVKWL NENAVEKVNI KPQVDRYRLK NGRRIILLAE GRLVNLGCAM GHPSFVMSNS FTNQVMAQIE LWTHPDKYPV GVHFLPKKLD EAVAEAHLGK LNVKLTKLTE KQAQYLGMSC DGPFKPDHYR Y | |
Greater than 90% as determined by SDS-PAGE. |
191 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |